siRNA Details
Gene Symbol: Ankrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107589.1 | NP_001101059.1 | 6 | 641 | ||
GenBank mRNA | FQ216123.1 | - | 6 | 651 | ||
FQ216298.1 | - | 6 | 651 | |||
FQ216609.1 | - | 6 | 653 | |||
FQ216805.1 | - | 6 | 651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
