siRNA Details
Gene Symbol: Loxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107592.1 | NP_001101062.1 | 12 | 1998 | ||
XM_006231393.2 | XP_006231455.1 | 12 | 2083 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Loxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107592.1 | NP_001101062.1 | 12 | 1998 | ||
XM_006231393.2 | XP_006231455.1 | 12 | 2083 |