siRNA Details
Gene Symbol: Cnnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107593.1 | NP_001101063.1 | 2, 3 | 960 | ||
XM_006231483.2 | XP_006231545.1 | 2, 3 | 1859 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_031396, AF202994, EF041508, DQ885890.
