siRNA Details
Gene Symbol: Cnnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107593.1 | NP_001101063.1 | 4 | 1178 | ||
XM_006231483.2 | XP_006231545.1 | 4 | 2077 | |||
XM_008760426.1 | XP_008758648.1 | 3 | 1936 |

More Information
Other Species Targeted

