siRNA Details
Gene Symbol: Tmem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107596.1 | NP_001101066.1 | 19, 20 | 3655 | ||
XM_006231163.1 | XP_006231225.1 | 20, 21 | 3704 | |||
XM_006231164.2 | XP_006231226.1 | 21, 22 | 3978 | |||
XM_006231165.2 | XP_006231227.1 | 21, 22 | 4816 | |||
XM_006231166.2 | XP_006231228.1 | 21, 22 | 3812 | |||
XM_008760298.1 | XP_008758520.1 | 22, 23 | 4153 | |||
XM_008760299.1 | XP_008758521.1 | 21, 22 | 3864 |
