siRNA Details
Gene Symbol: Tmem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107596.1 | NP_001101066.1 | 1 | 47 | ||
XM_006231163.1 | XP_006231225.1 | 1 | 60 | |||
XM_006231164.2 | XP_006231226.1 | 2 | 334 | |||
XM_006231165.2 | XP_006231227.1 | 2 | 1172 | |||
XM_006231166.2 | XP_006231228.1 | 2 | 168 | |||
XM_008760298.1 | XP_008758520.1 | 3 | 509 | |||
XM_008760299.1 | XP_008758521.1 | 2 | 220 |

More Information
Other Species Targeted

