siRNA Details
Gene Symbol: Dmrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107597.1 | NP_001101067.1 | 4 | 1826 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dmrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107597.1 | NP_001101067.1 | 4 | 1826 |