siRNA Details
Gene Symbol: Gbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191634.1 | NP_001178563.1 | 14 | 1822 | ||
XM_006231496.2 | XP_006231558.1 | 15 | 1998 | |||
XM_006231497.2 | XP_006231559.1 | 15 | 1995 | |||
XM_006231498.2 | XP_006231560.1 | 15 | 1998 | |||
XM_006231500.2 | XP_006231562.1 | 14 | 1899 | |||
XM_006231501.2 | XP_006231563.1 | 14 | 1899 | |||
XM_006231502.2 | XP_006231564.1 | 14 | 1896 | |||
GenBank mRNA | FQ216888.1 | - | 14 | 1878 |
