siRNA Details
Gene Symbol: Neurl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231510.2 | XP_006231572.1 | 4 | 1016 | ||
XM_006231511.2 | XP_006231573.1 | 4 | 873 | |||
XM_006231512.2 | XP_006231574.1 | 4 | 1416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
