siRNA Details
Gene Symbol: Kif20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107609.2 | NP_001101079.2 | 3 | 255 | ||
XM_006231281.2 | XP_006231343.1 | 3 | 307 | |||
XM_006231283.2 | XP_006231345.1 | 3 | 307 | |||
XM_006231284.2 | XP_006231346.1 | 3 | 307 | |||
XM_006231285.2 | XP_006231347.1 | 3 | 307 | |||
XM_008760328.1 | XP_008758550.1 | 3 | 464 | |||
XM_008760329.1 | XP_008758551.1 | 3 | 308 |

More Information
Other Species Targeted

Gene Ontology Categories:
