siRNA Details
Gene Symbol: Rbm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107611.2 | NP_001101081.2 | 2 | 1206 | ||
GenBank mRNA | EU562301.1 | - | 2 | 1206 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107611.2 | NP_001101081.2 | 2 | 1206 | ||
GenBank mRNA | EU562301.1 | - | 2 | 1206 |