siRNA Details
Gene Symbol: Med23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277054.1 | NP_001263983.1 | 28 | 3981 | ||
XM_008758612.1 | XP_008756834.1 | 29 | 4772 | |||
XM_008758613.1 | XP_008756835.1 | 27 | 4657 | |||
GenBank mRNA | BC090022.1 | - | 10 | 1643 |
