siRNA Details
Gene Symbol: Med23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277054.1 | NP_001263983.1 | 7 | 599 | ||
XM_008758612.1 | XP_008756834.1 | 7 | 1372 | |||
XM_008758613.1 | XP_008756835.1 | 7 | 1371 | |||
XM_008758614.1 | XP_008756836.1 | 7 | 1373 | |||
GenBank mRNA | FQ231601.1 | - | 7 | 581 |
