siRNA Details
Gene Symbol: Trim26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011665.2 | NP_001011665.1 | 6 | 1056 | ||
XM_008772692.1 | XP_008770914.1 | 7 | 1012 | |||
XM_008772693.1 | XP_008770915.1 | 5 | 2617 |
