siRNA Details
Gene Symbol: Trim39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213562.1 | NP_998727.1 | 7 | 1477 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178281, NM_024468, AK220298, AK166148, AB046382, and others.
Gene Ontology Categories:
