siRNA Details
Gene Symbol: RT1-CE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008832.2 | NP_001008832.1 | 5 | 928 | ||
XM_008772726.1 | XP_008770948.1 | 5 | 1059 | |||
XM_008772727.1 | XP_008770949.1 | 5 | 1062 | |||
GenBank mRNA | AJ005022.1 | - | 4 | 856 |
Gene Symbol: RT1-CE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008835.1 | NP_001008835.1 | 928 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
