siRNA Details
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212549.2 | NP_997714.2 | 4 | 519 | ||
GenBank mRNA | AB568263.1 | - | 3 | 303 | ||
BC098635.1 | - | 4 | 463 | 7098264 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212549.2 | NP_997714.2 | 4 | 519 | ||
GenBank mRNA | AB568263.1 | - | 3 | 303 | ||
BC098635.1 | - | 4 | 463 | 7098264 |