siRNA Details
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212491.2 | NP_997656.1 | 6 | 653 | ||
GenBank mRNA | BC085934.1 | - | 6 | 653 | 7112671 |

More Information
Other Species Targeted

Gene Ontology Categories:
