siRNA Details
Gene Symbol: Tmprss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107619.1 | NP_001101089.1 | 1 | 35 | ||
XM_006256210.2 | XP_006256272.1 | 2 | 125 | |||
XM_006256211.2 | XP_006256273.1 | 2 | 128 | |||
XM_008772817.1 | XP_008771039.1 | 1 | 517 |

More Information
Other Species Targeted

Gene Ontology Categories:
