siRNA Details
Gene Symbol: Ubash3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107620.1 | NP_001101090.1 | 13 | 2087 | ||
XM_006256215.2 | XP_006256277.1 | 14 | 2211 | |||
XM_008772818.1 | XP_008771040.1 | 14 | 1827 | |||
XM_008772819.1 | XP_008771041.1 | 14 | 1822 |
