siRNA Details
Gene Symbol: Ubash3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107620.1 | NP_001101090.1 | 8, 9 | 1512 | ||
XM_006256215.2 | XP_006256277.1 | 9, 10 | 1636 | |||
XM_008772818.1 | XP_008771040.1 | 9, 10 | 1252 | |||
XM_008772819.1 | XP_008771041.1 | 9, 10 | 1247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
