siRNA Details
Gene Symbol: Itgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037780.2 | NP_001032869.2 | 10 | 1391 | ||
GenBank mRNA | BC166721.1 | - | 10 | 1370 | 7932515 |

More Information
Other Species Targeted

Gene Ontology Categories:
