siRNA Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256367.2 | XP_006256429.1 | 11 | 1388 | ||
XM_008772883.1 | XP_008771105.1 | 11 | 2857 | |||
XM_008772884.1 | XP_008771106.1 | 10 | 1020 |

More Information
Other Species Targeted

Gene Ontology Categories:
