siRNA Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256367.2 | XP_006256429.1 | 11, 12 | 1465 | ||
XM_008772883.1 | XP_008771105.1 | 11, 12 | 2934 | |||
XM_008772884.1 | XP_008771106.1 | 10, 11 | 1097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
