siRNA Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107625.1 | NP_001101095.1 | 6 | 808 | ||
XM_006256367.2 | XP_006256429.1 | 6 | 794 | |||
XM_008772883.1 | XP_008771105.1 | 6 | 2263 | |||
XM_008772884.1 | XP_008771106.1 | 5 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
