siRNA Details
Gene Symbol: LOC103690413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772942.1 | XP_008771164.1 | 4 | 586 |

More Information
Other Species Targeted


UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC103690413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772942.1 | XP_008771164.1 | 4 | 586 |