siRNA Details
Gene Symbol: Herc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012074.1 | NP_001012074.1 | 26 | 3407 | ||
XM_006256134.2 | XP_006256196.1 | 25 | 3389 | |||
XM_006256135.2 | XP_006256197.1 | 25 | 3396 | |||
XM_006256136.2 | XP_006256198.1 | 24 | 3240 | |||
GenBank mRNA | BC087104.1 | - | 26 | 3407 | 7192894 | |
FQ218928.1 | - | 8 | 840 |
