siRNA Details
Gene Symbol: Herc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012074.1 | NP_001012074.1 | 4 | 677 | ||
XM_006256134.2 | XP_006256196.1 | 3 | 659 | |||
XM_006256135.2 | XP_006256197.1 | 4 | 690 | |||
XM_006256136.2 | XP_006256198.1 | 2 | 510 | |||
GenBank mRNA | BC087104.1 | - | 4 | 677 | 7192894 |
