siRNA Details
Gene Symbol: Vgll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107630.1 | NP_001101100.1 | 2 | 278 | ||
XM_006256448.2 | XP_006256510.1 | 2 | 274 | |||
XM_008772885.1 | XP_008771107.1 | 2 | 891 |
