siRNA Details
Gene Symbol: Gopc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107631.1 | NP_001101101.1 | 1 | 272 | ||
GenBank mRNA | BC168160.1 | - | 1 | 302 | 8364776 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gopc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107631.1 | NP_001101101.1 | 1 | 272 | ||
GenBank mRNA | BC168160.1 | - | 1 | 302 | 8364776 |