siRNA Details
Gene Symbol: Cdk19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107634.1 | NP_001101104.1 | 9 | 1142 | ||
XM_008772985.1 | XP_008771207.1 | 8 | 757 | |||
XM_008772986.1 | XP_008771208.1 | 5 | 438 |
