siRNA Details
Gene Symbol: RGD1310495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256539.2 | XP_006256601.1 | 2 | 243 | ||
GenBank mRNA | FQ210617.1 | - | 412 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1310495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256539.2 | XP_006256601.1 | 2 | 243 | ||
GenBank mRNA | FQ210617.1 | - | 412 |