siRNA Details
Gene Symbol: RGD1310495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256539.2 | XP_006256601.1 | 5 | 1288 | ||
GenBank mRNA | FQ210617.1 | - | 1603 |
Gene Symbol: RGD1304770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134547.1 | NP_001128019.1 | 4 | 1434 |
