siRNA Details
Gene Symbol: Fig4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047096.1 | NP_001040561.1 | 4, 5 | 446 | ||
XM_006256540.2 | XP_006256602.1 | 3, 4 | 1285 | |||
XM_006256541.2 | XP_006256603.1 | 2, 3 | 221 | |||
GenBank mRNA | DQ666853.1 | - | 4, 5 | 446 |
