siRNA Details
Gene Symbol: Scml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107638.1 | NP_001101108.1 | 8 | 1240 | ||
XM_006256572.2 | XP_006256634.1 | 8 | 1537 | |||
XM_008772989.1 | XP_008771211.1 | 4 | 1421 | |||
XM_008772990.1 | XP_008771212.1 | 4 | 837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
