siRNA Details
Gene Symbol: Sim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107641.1 | NP_001101111.1 | 11 | 2083 | ||
XM_006256604.2 | XP_006256666.1 | 12 | 2371 | |||
XM_008772994.1 | XP_008771216.1 | 12 | 2211 | |||
XM_008772995.1 | XP_008771217.1 | 12 | 3560 | |||
XM_008772996.1 | XP_008771218.1 | 12 | 2194 | |||
XM_008772997.1 | XP_008771219.1 | 11 | 5836 | |||
XM_008772998.1 | XP_008771220.1 | 9 | 1368 |
