siRNA Details
Gene Symbol: Phyhipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012076.1 | NP_001012076.1 | 5 | 866 | ||
XM_006256347.2 | XP_006256409.1 | 5 | 987 | |||
GenBank mRNA | BC078977.1 | - | 5 | 866 | 7111966 | |
FQ214387.1 | - | 1 | 52 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
