siRNA Details
Gene Symbol: Ap3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107646.1 | NP_001101116.1 | 22 | 2672 | ||
XM_006231814.2 | XP_006231876.1 | 24 | 4861 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ap3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107646.1 | NP_001101116.1 | 22 | 2672 | ||
XM_006231814.2 | XP_006231876.1 | 24 | 4861 |