siRNA Details
Gene Symbol: Xrcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006999.1 | NP_001007000.1 | 7 | 769 | ||
XM_006231751.1 | XP_006231813.1 | 7 | 938 | |||
XM_008760636.1 | XP_008758858.1 | 7 | 900 | |||
XM_008760637.1 | XP_008758859.1 | 7 | 965 | |||
XM_008760638.1 | XP_008758860.1 | 7 | 900 | |||
GenBank mRNA | BC083848.1 | - | 7 | 769 | 7122361 |
