siRNA Details
Gene Symbol: Ptcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107648.1 | NP_001101118.1 | 9 | 906 | ||
XM_006231847.2 | XP_006231909.1 | 10 | 977 | |||
XM_008760675.1 | XP_008758897.1 | 9 | 2167 |
