siRNA Details
Gene Symbol: Ddx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077647.1 | NP_001071115.1 | 17 | 1570 | ||
GenBank mRNA | GQ243745.1 | - | 16 | 1350 | ||
JQ013737.1 | - | 16 | 1358 | |||
S75275.1 | - | 17 | 1570 |

More Information
Other Species Targeted

Gene Ontology Categories:
