siRNA Details
Gene Symbol: Snx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107652.1 | NP_001101122.1 | 1, 2 | 1764 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_130796, AK077890, BC145951, BC063089, BC137801, and others.
Gene Ontology Categories:
