siRNA Details
Gene Symbol: RGD1306502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107654.1 | NP_001101124.1 | 6 | 1042 | ||
XM_006232047.2 | XP_006232109.1 | 6 | 1196 | |||
XM_006232048.2 | XP_006232110.1 | 6 | 984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
