siRNA Details
Gene Symbol: Drosha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107655.2 | NP_001101125.2 | 29 | 3714 | ||
XM_006232049.2 | XP_006232111.1 | 26 | 3537 | |||
XM_006232050.2 | XP_006232112.1 | 26 | 3513 | |||
GenBank mRNA | AY373464.1 | - | 2 | 226 | ||
FQ212690.1 | - | 8 | 648 | |||
FQ215440.1 | - | 9 | 708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
