siRNA Details
Gene Symbol: Drosha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107655.2 | NP_001101125.2 | 35 | 4261 | ||
XM_006232049.2 | XP_006232111.1 | 32 | 4084 | |||
XM_006232050.2 | XP_006232112.1 | 32 | 4060 | |||
GenBank mRNA | FQ212690.1 | - | 14 | 1195 | ||
FQ215440.1 | - | 15 | 1256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
