siRNA Details
Gene Symbol: Ankrd33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191565.1 | NP_001178494.1 | 3 | 550 | ||
GenBank mRNA | FQ232932.1 | - | 3 | 739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
