siRNA Details
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008770.3 | NP_001008770.1 | 6 | 633 | ||
XM_006232097.2 | XP_006232159.1 | 6 | 2744 | |||
GenBank mRNA | AY325197.1 | - | 6 | 635 | ||
BC088459.1 | - | 5 | 617 | 7127481 | ||
FQ209709.1 | - | 6 | 791 | |||
FQ213936.1 | - | 6 | 643 | |||
FQ215832.1 | - | 5 | 635 | |||
FQ219291.1 | - | 6 | 722 | |||
FQ219760.1 | - | 6 | 646 |

More Information
Other Species Targeted

Gene Ontology Categories:
