siRNA Details
Gene Symbol: Zfhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191702.1 | NP_001178631.1 | 2 | 594 | ||
XM_006232162.2 | XP_006232224.1 | 2 | 622 | |||
XM_006232164.2 | XP_006232226.1 | 2 | 487 | |||
XM_008760867.1 | XP_008759089.1 | 2 | 982 |
