siRNA Details
Gene Symbol: Samd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191703.1 | NP_001178632.1 | 4 | 353 | ||
XM_006232225.2 | XP_006232287.1 | 5 | 877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
