siRNA Details
Gene Symbol: Phc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107662.1 | NP_001101132.1 | 6 | 1631 | ||
XM_006232226.2 | XP_006232288.1 | 8 | 1805 | |||
XM_006232227.2 | XP_006232289.1 | 8 | 1805 | |||
XM_006232229.2 | XP_006232291.1 | 7 | 1706 | |||
XM_006232230.2 | XP_006232292.1 | 7 | 1706 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
